siRNA Details
Gene Symbol: 4930556N09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045358.1 | 1 | 35 | |||
GenBank mRNA | AK016152.1 | - | 1 | 36 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715755.1 | - | 1 | 36 | Chr. 10 - 97035768 - 97060926 [+] on Build GRCm38 |
