siRNA Details
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160318.1 | - | 1 | 536 | ||
BC112905.1 | - | 4895 | 3670722 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160318.1 | - | 1 | 536 | ||
BC112905.1 | - | 4895 | 3670722 |