siRNA Details
Gene Symbol: 1700022H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045488.1 | 4 | 459 | |||
GenBank mRNA | AK006240.1 | - | 4 | 460 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706323.1 | - | 4 | 460 | Chr. 12 - 9565236 - 9570585 [-] on Build GRCm38 |
