siRNA Details
Gene Symbol: Gm16283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079957.1 | - | 2 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB215047.2 | - | 2 | 511 | Chr. 18 - 49755794 - 49757186 [+] on Build GRCm38 |
