siRNA Details
Gene Symbol: Ctcflos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040321.1 | 3 | 2458 | |||
GenBank mRNA | AK005014.2 | - | 3 | 2459 | ||
AK079954.1 | - | 835 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctcflos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040321.1 | 3 | 2458 | |||
GenBank mRNA | AK005014.2 | - | 3 | 2459 | ||
AK079954.1 | - | 835 |