siRNA Details
Gene Symbol: Gm34907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079958.1 | - | 37 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB215187.2 | - | 2 | 48 | Chr. 14 - 103456372 - 103512616 [+] on Build GRCm38 |
