siRNA Details
Gene Symbol: C030014A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021077.1 | - | 1 | 204 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728157.1 | - | 1 | 201 | Chr. 13 - 117822316 - 117822978 [+] on Build GRCm38 |
