siRNA Details
Gene Symbol: 9130604C24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020298.1 | - | 1 | 928 | ||
BB546060.1 | - | 1 | 299 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130604C24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020298.1 | - | 1 | 928 | ||
BB546060.1 | - | 1 | 299 |