siRNA Details
Gene Symbol: 1700063H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006869.1 | - | 2 | 755 | ||
BG100370.1 | - | 1 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706854.1 | - | 2 | 755 | Chr. 6 - 122391379 - 122392687 [+] on Build GRCm38 |
