siRNA Details
Gene Symbol: 1700063A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040467.1 | 3 | 306 | |||
GenBank mRNA | AK006865.1 | - | 3 | 308 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706850.1 | - | 3 | 308 | Chr. 1 - 95990659 - 96021758 [-] on Build GRCm38 |
