siRNA Details
Gene Symbol: Gm38572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054526.1 | - | 4 | 608 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ053110.1 | - | 3 | 365 | Chr. 8 - 31601135 - 31663828 [-] on Build GRCm38 |
