siRNA Details
Gene Symbol: 4921529L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110484.1 | 2 | 201 | |||
GenBank mRNA | AK076597.1 | - | 2 | 201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614683.1 | - | 2 | 201 | Chr. 6 - 53978691 - 54016518 [+] on Build GRCm38 |
