siRNA Details
Gene Symbol: A430078I02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040219.1 | - | 114 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635659.1 | - | 1, 2 | 111 | Chr. 6 - 86472037 - 86480768 [-] on Build GRCm38 |
