siRNA Details
Gene Symbol: 4833428L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040732.1 | 2, 3 | 186 | |||
GenBank mRNA | AK014786.1 | - | 2, 3 | 188 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714279.1 | - | 2, 3 | 188 | Chr. 9 - 45417223 - 45431232 [-] on Build GRCm38 |
