siRNA Details
Gene Symbol: 4833428L15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040732.1 | 4, 5 | 495 | |||
GenBank mRNA | AK014786.1 | - | 4, 5 | 497 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714279.1 | - | 4, 5 | 497 | Chr. 9 - 45417223 - 45431232 [-] on Build GRCm38 |
