siRNA Details
Gene Symbol: 1700054A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045320.1 | 3 | 434 | |||
GenBank mRNA | AK018858.1 | - | 3 | 435 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706762.1 | - | 3 | 435 | Chr. 19 - 53076252 - 53084266 [+] on Build GRCm38 |
