siRNA Details
Gene Symbol: Gm10657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132138.1 | - | 1 | 947 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB321112.1 | - | 1 | 521 | Chr. 9 - 58555224 - 58555847 [+] on Build GRCm38 |
