siRNA Details
Gene Symbol: 1700052M18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018857.1 | - | 1 | 82 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706759.1 | - | 1 | 81 | Chr. 11 - 102664965 - 102665248 [-] on Build GRCm38 |
