siRNA Details
Gene Symbol: Tmem132cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038127.1 | 3 | 234 | |||
NR_038128.1 | 4 | 302 | ||||
NR_038129.1 | 4 | 444 | ||||
GenBank mRNA | AK007002.1 | - | 3 | 197 | ||
AK018855.1 | - | 3 | 235 | |||
AK018902.1 | - | 3 | 235 | |||
BY448567.1 | - | 3 | 236 |
