siRNA Details
Gene Symbol: Tmem132cos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038127.1 | 2, 3 | 160 | |||
NR_038128.1 | 3, 4 | 228 | ||||
GenBank mRNA | AK007002.1 | - | 2, 3 | 123 | ||
AK018855.1 | - | 2, 3 | 161 | |||
AK018902.1 | - | 2, 3 | 161 | |||
BY448567.1 | - | 2, 3 | 161 |
