siRNA Details
Gene Symbol: 3110038A09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014137.1 | - | 960 | |||
AV165602.1 | - | 1 | 237 | |||
AV165624.1 | - | 1 | 242 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK014161.1 | - | 547 | Chr. - - [-] on Build |
