siRNA Details
Gene Symbol: 2610024L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011538.1 | - | 1 | 195 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710795.1 | - | 1 | 195 | Chr. 6 - 120835382 - 120836074 [-] on Build GRCm38 |
