siRNA Details
Gene Symbol: 2610024J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011537.1 | - | 1 | 385 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710793.1 | - | 1 | 385 | Chr. 4 - 6941878 - 6942298 [-] on Build GRCm38 |
