siRNA Details
Gene Symbol: 4930511J24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015759.1 | - | 2 | 209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715302.1 | - | 2 | 209 | Chr. 12 - 109911785 - 109912707 [+] on Build GRCm38 |
