siRNA Details
Gene Symbol: Gm2233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC035543.1 | - | 2 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI739888.1 | - | 2 | 489 | Chr. 13 - 43948876 - 43951532 [+] on Build GRCm38 |
