siRNA Details
Gene Symbol: Snhg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034349.1 | - | 3386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB083265.2 | - | 1 | 658 | Chr. 7 - 59937374 - 59938054 [-] on Build GRCm38 |
