siRNA Details
Gene Symbol: Gm36017 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076918.1 | - | 4 | 892 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB015460.2 | - | 3 | 585 | Chr. 5 - 44412793 - 44426992 [-] on Build GRCm38 |
