siRNA Details
Gene Symbol: Gm30400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079520.1 | - | 2 | 1096 | ||
AK164315.1 | - | 3 | 418 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ064553.1 | - | 3 | 418 | Chr. 10 - 78747130 - 78765540 [+] on Build GRCm38 |
