siRNA Details
Gene Symbol: Gm30400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079520.1 | - | 3 | 2351 | ||
AK164315.1 | - | 5 | 1567 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB169021.2 | - | 1 | 523 | Chr. 10 - 78766514 - 78767155 [+] on Build GRCm38 |
