siRNA Details
Gene Symbol: 2900064K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013751.1 | - | 1 | 766 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712719.1 | - | 1 | 762 | Chr. 5 - 49518002 - 49518933 [-] on Build GRCm38 |
