siRNA Details
Gene Symbol: 4930540M03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040746.1 | 2 | 88 | |||
GenBank mRNA | AK019729.1 | - | 2 | 90 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715591.1 | - | 2 | 90 | Chr. 9 - 15619857 - 15641224 [+] on Build GRCm38 |
