siRNA Details
Gene Symbol: 4930538L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019723.1 | - | 2 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715567.1 | - | 2 | 252 | Chr. 14 - 47757034 - 47757498 [-] on Build GRCm38 |
