siRNA Details
Gene Symbol: 4930538E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019630.1 | - | 1 | 74 | ||
AK019722.1 | - | 1 | 31 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715563.1 | - | 1 | 31 | Chr. 11 - 24166873 - 24172781 [-] on Build GRCm38 |
