siRNA Details
Gene Symbol: 4930539C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019724.1 | - | 1 | 259 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715570.1 | - | 1 | 259 | Chr. 9 - 23749902 - 23750322 [+] on Build GRCm38 |
