siRNA Details
Gene Symbol: 4932412D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040521.1 | 4 | 602 | |||
GenBank mRNA | AK016521.1 | - | 4 | 603 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716203.1 | - | 4 | 603 | Chr. 16 - 42728930 - 42875587 [-] on Build GRCm38 |
