siRNA Details
Gene Symbol: C430039J01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021246.1 | - | 1 | 223 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY729179.1 | - | 1 | 223 | Chr. 7 - 116410141 - 116410803 [-] on Build GRCm38 |
