siRNA Details
Gene Symbol: 1700026F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045487.1 | 3 | 350 | |||
GenBank mRNA | AK006380.1 | - | 3 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706443.1 | - | 3 | 348 | Chr. 8 - 71006728 - 71026753 [-] on Build GRCm38 |
