siRNA Details
Gene Symbol: Gm13264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140051.1 | - | 3, 4 | 300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY727029.1 | - | 3, 4 | 300 | Chr. 2 - 11036874 - 11058780 [+] on Build GRCm38 |
