siRNA Details
Gene Symbol: Gm5817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135560.1 | - | 2 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB667280.1 | - | 1 | 96 | Chr. 17 - 86165471 - 86166842 [-] on Build GRCm38 |
