siRNA Details
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081556.1 | - | 1 | 2091 | ||
AK168156.1 | - | 1 | 776 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM946607.1 | - | 1 | 553 | Chr. 5 - 105612021 - 105612764 [+] on Build GRCm38 |
