siRNA Details
Gene Symbol: 5930436O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020044.1 | - | 1 | 994 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB629873.1 | - | 1 | 218 | Chr. 17 - 88057641 - 88058231 [-] on Build GRCm38 |
