siRNA Details
Gene Symbol: Bcas3os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046194.1 | 3 | 442 | |||
GenBank mRNA | AK020598.1 | - | 3 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627565.1 | - | 3 | 443 | Chr. 11 - 85766854 - 85775680 [-] on Build GRCm38 |
