siRNA Details
Gene Symbol: 4930547M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036704.1 | - | 2 | 1692 | ||
AK166420.1 | - | 2 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF449594.1 | - | 2 | 409 | Chr. 10 - 39899338 - 39907264 [+] on Build GRCm38 |
