siRNA Details
Gene Symbol: 2010205J10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008456.1 | - | 1 | 416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI605607.1 | - | 2 | 582 | Chr. 8 - 86063109 - 86063772 [+] on Build GRCm38 |
