siRNA Details
Gene Symbol: 4933429O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033783.1 | 3 | 336 | |||
GenBank mRNA | AK015198.1 | - | 3 | 292 | ||
AK016985.1 | - | 3 | 326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716669.1 | - | 3 | 326 | Chr. 14 - 48878884 - 48886191 [+] on Build GRCm38 |
