siRNA Details
Gene Symbol: Gm9962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033504.1 | 3 | 411 | |||
GenBank mRNA | AK049950.1 | - | 3 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB653224.1 | - | 3 | 412 | Chr. 7 - 57387272 - 57409168 [+] on Build GRCm38 |
