siRNA Details
Gene Symbol: F830002L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033558.1 | 3 | 1431 | |||
GenBank mRNA | AK158018.1 | - | 3 | 1431 | ||
AK162643.1 | - | 3 | 1434 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F830002L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033558.1 | 3 | 1431 | |||
GenBank mRNA | AK158018.1 | - | 3 | 1431 | ||
AK162643.1 | - | 3 | 1434 |