siRNA Details
Gene Symbol: 4930539C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040600.1 | 4 | 288 | |||
GenBank mRNA | AK015994.1 | - | 4 | 289 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715572.1 | - | 289 | Chr. - - [-] on Build |
