siRNA Details
Gene Symbol: 4930579D09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016315.1 | - | 2 | 257 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715954.1 | - | 2 | 260 | Chr. 6 - 143533963 - 143535510 [-] on Build GRCm38 |
