siRNA Details
Gene Symbol: 5830490A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018065.1 | - | 1 | 292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717998.1 | - | 1 | 292 | Chr. 10 - 64044072 - 64044910 [-] on Build GRCm38 |
